Learn More
Vernier Vernier Nitrogen Spectrum Tube
SDPSCI_ED
Catalog No. S16021ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Nitrogen Spectrum Tube
Catalog No. Product Type
S16021ND Nitrogen Spectrum Tube
1 options
Catalog No. S16021ND Supplier Vernier Supplier No. STN
Add to Cart
Add to Cart
Gas tubes for qualitative study by students noting more spectral lines and unique characteristic lines. For use with the Spectrum Tube Single Power Supply and the Spectrum Tube Carousel Power Supply.
  • Permanently enclosed in plastic carriers to prevent breakage
  • No through-the-glass electrodes
  • Ultrasafe
  • Long lasting and superior to older designs
  • For Grades 9 and up
Specifications
For Use With (Equipment) Spectrum Tube Single Power Supply and the Spectrum Tube Carousel Power Supply
Material Plastic
Product Type Nitrogen Spectrum Tube
WARNING: Cancer - www.P65Warnings.ca.gov