Learn More
Vernier Vernier pH Storage Solution Bottles
SDPSCI_ED
Catalog No. S16107ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Vernier pH Sensors
Catalog No. For Use With (Equipment)
S16107ND Vernier pH Sensors
1 options
Catalog No. S16107ND Supplier Vernier Supplier No. BTL
Add to Cart
Add to Cart
For use with Vernier pH Sensors, Tris-Compatible Flat pH Sensors, and Go Wireless™ pH.
  • The same bottles in which Vernier pH Sensors are shipped
  • Use them to make mini-beakers for microscale titrations, or to simply replace the storage bottles
Specifications
For Use With (Equipment) Vernier pH Sensors
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov