Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Primary Productivity Kit
SDPSCI_ED

Catalog No. S15971ND Shop All Vernier Software Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S15971ND 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15971ND Supplier Vernier Supplier No. PPK
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • For use with Dissolved Oxygen Probe (S66187) and Vernier Optical DO Probe (S16228ND)
  • Kit is an accessory for the Primary Productivity Lab
  • Consists of a box of 7 plastic bottles, 7 rubber stoppers, and a set of screens

Specifications

Grade 9 to Undergraduate
Includes 7 plastic bottles, 7 rubber stoppers, and a set of screens
Product Type Primary productivity Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.