Learn More
Vernier Vernier Respiration Monitor Belt
SDPSCI_ED
Catalog No. S16070ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Gas Pressure Sensor
Catalog No. For Use With (Equipment)
S16070ND Gas Pressure Sensor
1 options
This item is intended for educational purposes only.
Catalog No. S16070ND Supplier Vernier Supplier No. RMB
This item is intended for educational purposes only.
Requires
Requires a Vernier Gas Pressure Sensor, a compatible interface, and software to collect data. (All sold separately)
Used with the Gas Pressure Sensor to measure human respiration rates.
  • Simply strap the belt around the chest and pump air into belt with hand bulb
  • Monitor the pressure associated with the expansion and contraction of chest during breathing
  • Recommended for all grades
  • Compatible Interfaces include: LabQuest™ 2, Original LabQuest, LabQuest Mini, GoLink™, SensorDAQ™, NXT Sensor Adapter™, LabPro™, EasyLink™, TI-Nspire Lab Cradle™
Order Info
Gas Pressure Sensor, interface, and software sold separately.
Specifications
For Use With (Equipment) Gas Pressure Sensor
Product Type Respiration Monitor Belt
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov