Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier A Den of Inquiry, Volume 1
SDPSCI_ED

Catalog No. S16140ND Shop All Vernier Software Products
Change view
Click to view available options
15 lab activities for introductory physics
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16140ND 15 lab activities for introductory physics
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16140ND Supplier Vernier Supplier No. DEN

Collection of lab activities for introductory physics.

  • Contains 15 lab activites
  • Pay attention to data analysis and mathematical modeling without using sensors
  • Includes teacher notes and blackline masters for students predictions, instructions for lab and for data analysis, and follow-up questions

Specifications

For Use With (Application) For learning introductory physics
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.