Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier A Den of Inquiry Volume 2
SDPSCI_ED

Catalog No. S15960ND Shop All Vernier Software Products
Change view
Click to view available options
Collection of activities for introductory physics
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15960ND Collection of activities for introductory physics
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15960ND Supplier Vernier Supplier No. DEN2

missing translation for 'validMainframeMsg'

Collection of activities for introductory physics.

  • Contains 16 sensor-based activities
  • Topics include kinematics, mechanics and energy; activities emphasize data analysis and modeling
  • Either Fathom 2.1 or Logger Pro software can be used
  • Each lab includes blackline masters for student handouts and extensive teacher notes
  • Includes extensions and options for making the activities more open-ended

Specifications

For Use With (Application) For learning basic physics
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.