Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Advanced Physics with Vernier - Mechanics
SDPSCI_ED

Catalog No. S15966ND Shop All Vernier Software Products
Change view
Click to view available options
Advanced Mechanical Physics with Vernier probeware
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15966ND Advanced Mechanical Physics with Vernier probeware
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15966ND Supplier Vernier Supplier No. PHYSAM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Learning mechanics concepts using Vernier probeware.

  • For in-depth introductory physics course, such as college physics, AP Physics, or IB Physics
  • Designed for interactive teaching style, with planned notes for instructor or student-led discussion
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) Learning concepts of mechanics using Vernier probeware
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.