Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Chemistry with Vernier
SDPSCI_ED

Catalog No. S16047ND Shop All Vernier Software Products
Change view
Click to view available options
Contains more than 36 chemistry-related experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16047ND Contains more than 36 chemistry-related experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16047ND Supplier Vernier Supplier No. CWV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Foundational chemistry book for high school and college courses.

  • Contains more than 36 experiments in thermochemistry, gas laws, acid-base reactions, etc
  • Experiments included for Gas Pressure Sensor, Temperature Probe, pH Sensor, Conductivity Probe, Colorimeter, Voltage Probe
  • Includes materials list, step-by-step instructions, data tables, and questions
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD
  • Recommended for College and High School

Specifications

For Use With (Application) Experiments in thermochemistry, electrochemistry, acid-base reactions
Product Type Lab Book
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.