Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Earth Science with Vernier
SDPSCI_ED

Catalog No. S15989ND Shop All Vernier Software Products
Change view
Click to view available options
Contains 33 Earth Science experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15989ND Contains 33 Earth Science experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15989ND Supplier Vernier Supplier No. ESV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For learning Earth Science, geology, etc

  • Contains 33 experiments on geology, soil analysis, water quality, hydrology, oceanography, meteorology, and energy
  • Recommended for College, High School, and Middle School Science
  • Includes materials list, step-by-step instructions, data tables, and questions
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) For learning Earth Science topics
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.