Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Human Physiology with Vernier
SDPSCI_ED

Catalog No. S16036ND Shop All Vernier Software Products
Change view
Click to view available options
Contains 24 experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16036ND Contains 24 experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16036ND Supplier Vernier Supplier No. HPA
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Activities detailing about the physiology of human organ systems.

  • Contains 24 experiments
  • Include measuring activity of upper respiratory tract, lungs, heart, nervous system, and musculoskeletal system
  • Illustrate response of human body to stress or external influences, with homeostasis as underlying theme
  • Recommended for College and High School
  • Includes materials list, step-by-step instructions, data tables, and questions
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) For learning physiology of human organs
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.