Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Investigating Biology through Inquiry
SDPSCI_ED

Catalog No. S16097ND Shop All Vernier Software Products
Change view
Click to view available options
Investigating Biology through Inquiry
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16097ND Investigating Biology through Inquiry
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16097ND Supplier Vernier Supplier No. BIOI
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Helps integrate inquiry into biology curriculum.

  • Contains investigations for many fundamental concepts in biology
  • Includes preliminary activity, teacher information, sample researchable questions, and sample data
  • Labs correlate to AP and IB standards

Specifications

For Use With (Application) Investigations for many fundamental concepts in biology
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.