Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Lets Go Investigating Temperature
SDPSCI_ED

Catalog No. S16143ND Shop All Vernier Software Products
Change view
Click to view available options
Collection of 10 temperature experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16143ND Collection of 10 temperature experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16143ND Supplier Vernier Supplier No. ELBTEMP
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Collection of engaging temperature experiments.

  • Contains 10 activities
  • For grade 2 through 5
  • Easy-to-use, black-line masters with illustrations and teacher tips

Specifications

For Use With (Application) For learning temperature concepts
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.