Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Middle School Science with Vernier
SDPSCI_ED

Catalog No. S15962ND Shop All Vernier Software Products
Change view
Click to view available options
Contains 38 experiments in Earth science, life science, and physical science
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S15962ND Contains 38 experiments in Earth science, life science, and physical science
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S15962ND Supplier Vernier Supplier No. MSV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains experiment activities in Earth science, life science and physical science.

  • For students in grades 6-8
  • Contains 38 experiments using 10 different Vernier middle school sensors
  • Contains preliminary activities with step-by-step instructions, data tables, and questions
  • Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD

Specifications

For Use With (Application) 38 experiments in middle school science
Product Type Lab Book
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.