Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Physical Science through Inquiry
SDPSCI_ED

Catalog No. S16057ND Shop All Vernier Software Products
Change view
Click to view available options
Contains 40 experiments on physical science
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16057ND Contains 40 experiments on physical science
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16057ND Supplier Vernier Supplier No. PSV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For learning introductory physical science and integrated science classes

  • Contains 40 ready-to-use experiments
  • Experiments included for 9 Vernier physical science sensors in chemistry and physics
  • Includes materials list, step-by-step instructions, data tables, and questions
  • Contains Teacher Information section with directions for setting up experiments, helpful hints, and sample graphs and data
  • Word-processing files of student sections on CD
  • Recommended for High School and K-8

Specifications

For Use With (Application) For introductory physical science and integrated science classes
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.