Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Chemistry Investigations for Use with AP Chemistry
SDPSCI_ED

Catalog No. S16079ND Shop All Vernier Software Products
Change view
Click to view available options
16 inquiry-based laboratory experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16079ND 16 inquiry-based laboratory experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16079ND Supplier Vernier Supplier No. APCHEM
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Guides students to investigate key principles of AP Chemistry using electronic sensors to collect important data.

  • Contains 16 inquiry-based lab experiments
  • Includes sample data, sample analysis and calculations
  • Guide to preparing reagents and chemical solutions
  • Includes step-by-step instructions for operating probes and software

Specifications

For Use With (Application) Chemistry investigations for use with AP chemistry
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.