Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Software and Technology Water Quality with Vernier
SDPSCI_ED

Catalog No. S16289ND Shop All Vernier Software Products
Change view
Click to view available options
Contains 18 water quality tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S16289ND Contains 18 water quality tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16289ND Supplier Vernier Supplier No. WQV
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes 18 tests including pH, total dissolved solids, dissolved oxygen, BOD, flow rate, turbidity, nitrates, and phosphates

  • Includes water testing
  • Supports nine tests in the Water Quality Index (WQI)
  • Comprehensive introduction included for each test

Specifications

For Use With (Application) Contains 18 water quality tests
Product Type Lab Book
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.