Learn More
Vernier Vernier Spectrum Tube Single Power Supply
SDPSCI_ED
Catalog No. S16017ND Shop All Vernier Software Products
Change view
Click to view available options
Product Type:
Spectrum Tube Single Power Supply
Catalog No. Product Type
S16017ND Spectrum Tube Single Power Supply
1 options
Catalog No. S16017ND Supplier Vernier Supplier No. STSPS
Add to Cart
Add to Cart
Energizes a single tube to simply view gas discharge spectral lines with any of Venier spectrometers.
  • Ultra-safe
  • No exposed high voltage
  • Energize one tube at a time
  • Storage space for an additional six tubes
  • Grades 9 and up
  • Includes Spectrum Tube Fiber Optic Holder
  • Gas spectrum tubes sold separately
Specifications
Includes Spectrum Tube Single Power Supply, Spectrum Tube Fiber Optic Holder
Product Type Spectrum Tube Single Power Supply
WARNING: Cancer - www.P65Warnings.ca.gov