Learn More
Vernier Standard High Cl ISE Solution
SDPSCI_ED
Catalog No. S16123ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
High Chloride ISE
Catalog No. For Use With (Equipment)
S16123ND High Chloride ISE
1 options
Catalog No. S16123ND Supplier Vernier Supplier No. CLHST
Add to Cart
Add to Cart
Used to calibrate Ion-Selective Electrodes
  • 500mL solution has concentration of 1000mg/L
Specifications
For Use With (Equipment) High Chloride ISE