Learn More
Vernier Standard High Nitrate ISE Solution
SDPSCI_ED
Catalog No. S16218ND Shop All Vernier Software Products
Catalog No. S16218ND Supplier Vernier Supplier No. NO3HST
Add to Cart
Add to Cart
Used to calibrate Ion-Selective Electrodes
  • 500mL solution has concentration of 100mg/L
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov