Learn More
Vernier Standard Low Ammonium ISE Solution
SDPSCI_ED
Catalog No. S16216ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Equipment):
Low ammonium ISE
Catalog No. For Use With (Equipment)
S16216ND Low ammonium ISE
1 options
Catalog No. S16216ND Supplier Vernier Supplier No. NH4LST
Add to Cart
Add to Cart
Used to calibrate Ion-Selective Electrodes
  • 500mL solution has concentration of 1mg/L
Specifications
For Use With (Equipment) Low ammonium ISE
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov