Learn More
Vernier Standard Low Potassium ISE Solution
SDPSCI_ED
Catalog No. S16178ND Shop All Vernier Software Products
Catalog No. S16178ND Supplier Vernier Supplier No. KLST
Add to Cart
Add to Cart
Used to calibrate Ion-Selective Electrodes
  • 500mL solution has concentration of 10mg/L
WARNING: Cancer - www.P65Warnings.ca.gov