Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vernier Vernier UV-VIS Spectrophotometer Optical Fiber
SDPSCI_ED

Catalog No. S16286ND Shop All Vernier Software Products
Change view
Click to view available options
For Use With (Application):
Emission spectrum experiments
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
S16286ND Emission spectrum experiments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S16286ND Supplier Vernier Supplier No. VSPFIBER
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For emission spectrum experiments.

Designed exclusively for use with the Vernier UV-VIS Spectrophotometer.
  • Simply insert unit into cuvette holder and point tip of 1m optical fiber at a light source
  • Unit is keyed for safety
  • Wavelength range: from 350 to 900nm

Specifications

For Use With (Application) Emission spectrum experiments
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.