Learn More
ATCC Cercopithecus aethiops
Supplier:  ATCC CCL81/DEL
Encompass
The Vero cell line was initiated in 1962 from the kidney tissue derived from a normal, adult African green monkey. The cell line can be used in a variety of applications, including the detection of verotoxins, detection of virus in ground beef, efficacy testing, the study of malaria, media testing, vaccine development, protein expression, and mycoplasma testing. The Vero cell line is also a suitable transfection host.
Catalog No. 50-188-381FP