Learn More
ATCC Vero, Select Kidney; Monkey (Chlorocebus aethiops) Grivet
Catalog No. 502382558
Encompass
The Vero Select cell line was derived from Vero, CCL-81. The cell line can be used in a variety of applications, including the detection of verotoxins, detection of virus in ground beef, efficacy testing, the study of malaria, media testing, vaccine development, protein expression, and mycoplasma testing. The Vero cell line is also a suitable transfection host.
Catalog No. 50-238-2558 Supplier ATCC Supplier No. CCL81.2
The Vero Select cell line was derived from Vero, CCL-81. The cell line can be used in a variety of applications, including the detection of verotoxins, detection of virus in ground beef, efficacy testing, the study of malaria, media testing, vaccine development, protein expression, and mycoplasma testing. The Vero cell line is also a suitable transfection host.