Learn More
ATCC Vero STAT1 KO; CRISPR/Cas9 Mod. Cell Line; African Green Monkey kidney; (Chlorocebus aethiops) Grivet
Catalog No. 502382544
Encompass
Vero.STAT1 KO is a CRISPR/Cas9 genome-edited STAT1 knockout cell line that exhibits significantly increased virus production capacity for viral vaccine manufacturing.
Catalog No. 50-238-2544 Supplier ATCC Supplier No. CCL81VHG
Add to Cart
Add to Cart
Request a Quote
Vero.STAT1 KO is a CRISPR/Cas9 genome-edited STAT1 knockout cell line that exhibits significantly increased virus production capacity for viral vaccine manufacturing.