Learn More
TARGETMOL CHEMICALS INC VEROFYLLINE 5MG
SDP
Catalog No. 502580236
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Verofylline (Verofyllinum) is an orally available long-acting multiacting methylxanthine-substituted bronchodilator with inhibitory effects on PDE4 for the treatment of asthma disease research obesity. purity: 99%
Catalog No. 50-258-0236 Supplier TARGETMOL CHEMICALS INC Supplier No. T350485MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. Verofylline (Verofyllinum) is an orally available long-acting multiacting methylxanthine-substituted bronchodilator with inhibitory effects on PDE4 for the treatment of asthma disease research obesity. purity: 99%