Learn More
AdipoGen Verruculogen
SDP
Supplier:  AdipoGen BVT0443M001
Encompass_Preferred
Chemical. CAS 12771-72-1. Formula C27H33N3O7. MW 511.6. Isolated from Aspergillus fumigatus. Fumitremorgin-type alkaloid. Tremorgenic neurotoxic mycotoxin. Selective blocker of KCa1.1 high-conductance Ca2-activated K BK maxi-K channels. Decreases GABA levels in CNS. Increases spontaneous glutamate and aspartate release. At toxic doses, increases the number and decreases the affinity of DHP receptors in rat cortex. Cell cycle inhibitor at the M phase. Inhibits calmodulin and PKC activity. Has apoptotic effects on human T lymphocytes. Stimulates the growth of seedling roots of buckwheat.
Catalog No. 50-256-1922