Learn More
Enzo Life Sciences Verruculogen (5mg). CAS: 12771-72-1
SDP
Catalog No. 502008934
Encompass_Preferred
Neurotoxin that acts as an inhibitor of the M phase of the mammalian cell cycle. Inhibits Ca2+-activated K+ channels. Decreases GABA levels in CNS. Purity: ≥95% (HPLC, TLC). Appearance: White to off-white powder. Solubility: Soluble in ethyl acetate, DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.
Catalog No. 50-200-8934 Supplier Enzo Life Sciences Supplier No. ALX380056M005
Neurotoxin that acts as an inhibitor of the M phase of the mammalian cell cycle. Inhibits Ca2+-activated K+ channels. Decreases GABA levels in CNS. Purity: ≥95% (HPLC, TLC). Appearance: White to off-white powder. Solubility: Soluble in ethyl acetate, DMSO, 100% ethanol or methanol. Long Term Storage: -20°C.