Learn More
Fisherbrand™ Versa-Clean™ Pharmaceutical Production and Multi-Purpose Cleaner
Shop All Fisherbrand Products

Description
- Clean-out-of-place (COP): Manual labware washing, pharmaceutical washers, immersion soak tanks, ultrasonic cleaners. Clean-in-place (CIP): Mixing vessels, process equipment, piping, packaging equipment
- Use where glassware and sensitive lab equipment must be totally free of residues that could interfere with analysis, tissue culture, manufacturing and other procedures
- Third party lot specific tested for inhibitory residues to meet NELAC and State water lab audit criteria (Standard Methods for the Examination of Water and Wastewater 21st Ed., 9020B-4A-2)
- Does not contain: alcohol, ammonia, APE’s, caustic , chlorine, fragrance, glycol ether solvents, NPE’s, NTA, phosphates, silicates, solvents, sulfates, volatile organic compounds or ozone depleting substances
- Readily biodegradable in 7 days (OECD 301D) and phosphate free
- Mild pH of 9.75 it can be disposed of straight to the drain
- Complete solubility, works equally well in hot or cold water
- Formulated for controlled foaming
- Fisherbrand Versa-Clean is manufactured using sustainable, low waste and low energy processes in an audited, modern plant, using state of the art, precision production equipment
Certifications
Passes inhibitory residue test.
Specifications
Specifications
| Form | Liquid |
| For Use With (Application) | For use in ultrasonic cleaners, high-pressure sprayers, scrubbers/soak tanks |
| Volume (Metric) | 4 L |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.