Learn More
Chemglass Life Sciences Versa-Roll Drum Only, 16-18 mm Tubes, 64-Position (For Versa-Roll Roller)
SDP
Catalog No. NC3901950
Encompass
Drum-only accessory for the Chemglass Versa-Roll digital tube roller. The removable drum provides 64 positions and accommodates tubes with diameters between 16 and 18 mm, allowing users to configure the roller for small-diameter culture or sample tubes.
  • Removable 64-position drum for small-diameter tubes.
  • Holds tubes 16-18 mm in diameter.
  • Enables rapid changeover between tube sizes to increase workflow flexibility.
  • Compatible with the matching digital tube roller base.
  • Designed for laboratory use and routine sample processing.
Catalog No. NC3901950 Supplier Chemglass Life Sciences Supplier No. CLS3869A03
Drum-only accessory for the Chemglass Versa-Roll digital tube roller. The removable drum provides 64 positions and accommodates tubes with diameters between 16 and 18 mm, allowing users to configure the roller for small-diameter culture or sample tubes.
  • Removable 64-position drum for small-diameter tubes.
  • Holds tubes 16-18 mm in diameter.
  • Enables rapid changeover between tube sizes to increase workflow flexibility.
  • Compatible with the matching digital tube roller base.
  • Designed for laboratory use and routine sample processing.