Learn More
Bellco Glass, Inc. VersaBarb, Spigot Fitting, 1 1/8 Thread, 3/4" Sanitary Connect, 1/EA
SDP
Supplier:  Bellco Glass, Inc. 452800420
Encompass_Preferred
VersaBarb provides flexibility in drainage, pump connection, filling bell, and automated equipment. VersaBarb is available in 1/4", 3/8", 1/2", and 3/4" hose barb sizes. In addition, a 3/4" sanitary fitting VersaBarb is also available to enable the EZgrip carboy to function as a supply reservoir for a larger system.Offers a convenient solution for bottom-connections and can fit a variety of tubing diametersMade from Class VI autoclavable polypropyleneAvailable in 1/4", 3/8", 1/2", and 3/4" hose barb sizes or 3/4" sanitary fitting*Only compatible with EZgrip and EZBio Carboys. Thread: 1-1/8Material: PolypropyleneHeight: 2.1" (54 mm)Diameter: 1.5" (38 mm)
Catalog No. 50-255-0342