Learn More
BEST SANITIZERS AD10048B VersaClenz Touchless Dispenser – Black Each All VersaClenz Compatible Cartridges N/A 2 7x12x4.5
Supplier:  BEST SANITIZERS AD10048B
Encompass_Preferred
Reduce cross-contamination with the VersaClenz Touchless Dispenser. This versatile system can dispense different products including sprays, foams, liquids, lotions, soaps and sanitizers. Using the same universal 1000mL and 1250mL cartridge as its manual counterpart, the VersaClenz Touchless system allows for drastic reduction of SKUs and warehousing needs without any reduction in product options. Available in black or white. Automatic dispensers recommended for dry placement areas only.
Catalog No. NC2953647