Learn More
3M™ Versaflo™ Back Pack Adaptor for TR-600/800 PAPR
Catalog No. 19072000 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-072-000 1 Ea.
1 options
Catalog No. 19-072-000 Supplier 3M™ Supplier No. 7100035843
Add to Cart
Add to Cart
Enables the TR-600 series PAPR to be worn when paired with the BPK-01 backpack.
  • Securely holds TR-600 in place
  • Multi-layer construction secures TR-600 in place. The TR-655 back pack adapter, enables the TR-600 series PAPR to be worn on the BPK-01 backpack
  • This configuration allows the TR-600 series PAPR to be worn on the wearer's back rather than on a belt at the waist
  • 3M Catalog# TR-655/37385
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance NIOSH Approved
Disposable Reusable
Product Type Backpack Adapter
For Use With (Equipment) TR-600 and TR-800 PAPR Systems
Model TR-655
Quantity 1 Ea.