Learn More
3M™ Versaflo™ Battery Adapter
Catalog No. 19071194 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Breathe Easy™, GVP Systems
Catalog No. For Use With (Equipment)
19-071-194 Breathe Easy™, GVP Systems
1 options
Catalog No. 19-071-194 Supplier 3M™ Supplier No. 7100127544
Add to Cart
Add to Cart
Battery adapter allows use of the TR-630 battery with GVP PAPR systems.
  • Easy to use
  • Replaces BP-15 battery for certain GVP and Breathe Easy RBE & FR Systems
  • Works with 3M™ Standard Battery TR-630
  • 3M Catalog# TR-659
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Breathe Easy™, GVP Systems