Learn More
3M™ Versaflo™ Cleaning and Storage Kit BT-953
Catalog No. 19070959 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
19-070-959 1 Ea.
1 options
Catalog No. 19-070-959 Supplier 3M™ Supplier No. 7100176699
  • $79.00
    $68.65 / Case of 1
    Save $10.35
    13% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Includes two caps and one plug for the openings of the BT-63/64 where they would connect to a facepiece and respirator. Designed to assist with cleaning and storage for the BT-63/64 breathing tubes.
  • Enables BT-63/64 Breathing Tubes to be submerged for cleaning
  • Helps protect BT-63/64 Breathing Tubes from internal contamination during cleaning
  • Helps protect BT-63/64 Breathing Tubes from internal contamination during storage
  • 3M Catalog# BT-953
TRUSTED_SUSTAINABILITY
Specifications
Model BT-953
For Use With (Equipment) Versaflo™ Powered Air Purifying Respirators
Quantity 1 Ea.