Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Easy Clean Holster

Catalog No. 19071193 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-071-193 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-071-193 Supplier 3M™ Supplier No. 7100127542
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PAPR holster is designed to keep the battery adapter and battery secure and protected during use. For use with TR-659 adapter and TR-630 battery.

  • Easy Clean
  • Provides protection and stability for 3M™ Versaflo™ Battery Adapter TR-659
  • 3M Catalog# TR-657

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Approved
Color Black
Disposable Reusable
Product Type Holster
For Use With (Application) Nuclear Energy, Pharmaceutical, Research, and Production
For Use With (Equipment) TR-659 Adapter and TR-630 Battery
Model TR-657
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.