Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Headcover PAPR Kit

Catalog No. 19121502 Shop All 3M Products
Change view
Click to view available options
Size:
Large/X-Large
Small/Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-121-502 Large/X-Large
19-121-501 Small/Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-121-502 Supplier 3M™ Supplier No. 7100098538
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Headcover PAPR kit offers a complete powered air respirator system that includes all necessary components to get started.

  • Cover your bases - Utilizes cartridges to protect against gas and organic vapors, combined with filters for protection against particulates.
  • Get more out of your workday - Longer run time, reduced downtime with a high capacity battery and charge-level indicators to keep you prepared and informed.
  • A broader view - See more of your work world with this convenient, maintenance-free disposable headcover.
  • Keeping it clean - Fully submersible product features a smooth surface design that’s quick and easy to clean and decontaminate.
  • Gas and Vapor protection can be provided with cartridges not part of this kit
  • System provides integrated solutions for respiratory protection, with limited eye, head, face and skin protection as well
  • Includes: Headcover, TR-602N PAPR Unit, TR-630 Standard Battery, BT-30 Length Adjusting Breathing Tube, TR-627 Easy Clean Belt, TR-971 Airflow Indicator, TR-6712N (HE) Particulate Filter, TR-6700FC Filter Cover, TR-641N Single Station Battery Charger, TR-653 Cleaning and Storage Kit, TR-991 Storage Bag
  • 3M Catalog# TR-600-HKL , 3M Catalog# TR-600-HKS

TRUSTED_SUSTAINABILITY

Specifications

Battery Life 4 to 12 hours
Battery Type Lithium-ion
Cartridge Type Gas and Organic Vapors (sold separately)
Certifications/Compliance NIOSH Approved
Color Gray
Filter Type HEPA
Product Type PAPR Healthcare Systems and Kits
Size Large/X-Large
For Use With (Application) Pharmaceutical and Healthcare
Model TR-600-HKL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.