Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Heavy Duty Tight Fitting Breathing Tube

Catalog No. 19070957 Shop All 3M Products
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-070-957 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-957 Supplier 3M™ Supplier No. 7100167214
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Connects to 3M™ full facepiece tight fitting respirator to Versaflo™ TR-600 PAPR.

  • Heat-resistant neoprene rubber material for rugged applications
  • Connects full facepiece tight fitting respirator to a 3M™ Versaflo™ TR-600 PAPR
  • Fully submersible for cleaning when using cleaning and storage kit
  • 3M Catalog# BT-64

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance NIOSH Approved
Color Black
Product Type Breathing Tube
For Use With (Application) General Maintenance, spray coating, demolition, liquid chemical handling
For Use With (Equipment) Versaflo™ PAPR Systems
Includes BT-64 Heavy Duty Breathing Tube
Model BT-64
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.