Learn More
MSC VERSAFLO HLTHCAR PAPR KIT
Catalog No. 5000688292
Encompass
This PAPR and SAR complete system is designed for use in pharmaceutical environments, specifically for powder handling applications. It features a large size and utilizes a reliable lithium-ion battery.
  • PAPR system type
  • Ideal for powder handling in pharmaceuticals
  • Lithium-ion battery chemistry
  • Large size
Catalog No. 50-006-88292 Supplier MSC Supplier No. 90259789
This PAPR and SAR complete system is designed for use in pharmaceutical environments, specifically for powder handling applications. It features a large size and utilizes a reliable lithium-ion battery.
  • PAPR system type
  • Ideal for powder handling in pharmaceuticals
  • Lithium-ion battery chemistry
  • Large size