Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-930 Visor Lock Kit

Catalog No. 19071139 Shop All 3M Products
Change view
Click to view available options
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-071-139 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-071-139 Supplier 3M™ Supplier No. 7100198700
Add to Cart
Add to Cart
This item is not returnable. View return policy

Visor lock kit is an accessory to Versaflo™ M-400 Series helmets.

  • Restricts lifting the visor frame of an M-400 series helmet, helping to prevent lifting of the visor when preferred/needed for use.
  • Lock mechanically attaches the visor frame to the jaw of M-400 series helmets with the intent to prevent lifting of the visor if needed or preferred by users.
  • Sold as a retrofit kit with 5 locks
  • Can be installed on new or existing helmets.
  • Helmet purchased separately.
  • Locks M-400 series faceshield in the closed position.
  • For use with all M-400 series helmets, including the M-401, M-405, M-407, M-407SG and M-409SG.
TRUSTED_SUSTAINABILITY

Specifications

Color Silver
Disposable Reusable
Material Metal
Product Type Visor Locking Kit
For Use With (Application) Remediation
For Use With (Equipment) M-400 Series Helmets, M-401, M-405, M-407, M-407 SG, M-409 SG
Includes 5 Visor Locks, 10 screws, Drill Guide
Model M-930
Quantity 5/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.