Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-Series Helmet Cover

Catalog No. 19070934 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo™ M-Series Helmet Assembly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-070-934 Versaflo™ M-Series Helmet Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-934 Supplier 3M™ Supplier No. 7100168652
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Designed to provide additional protection to both the wearer and the helmet. Compatible with Versaflo™ M-400 Series systems.

  • Compatible with M-400 helmets
  • Available as replacement part
  • Designed to provide additional protection to both the wearer and the safety helmet
  • Optional shroud for use with Versaflo™ M-407SG helmets featuring L-153SG Speedglas™ attachment
  • 3M Catalog# M-978

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI Z89.1 Type I, Class G
Color Black
Disposable Reusable
Product Type Helmet Cover
Size Standard
For Use With (Application) Welding
For Use With (Equipment) Versaflo™ M-Series Helmet Assembly
Includes Helmet Cover Only
Model M-978
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.