Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ M-Series Leather Shroud

Catalog No. 19070933 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
M-Series Helmet Assembly
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-070-933 M-Series Helmet Assembly
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-933 Supplier 3M™ Supplier No. 7100289614
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Leather shroud for use with Versaflo™ M-400 Series helmets.

  • Contoured brow guard rests comfortably against forehead and helps provide additional coverage from flying particles
  • Leather shroud compatible only with existing Versaflo™ M-400 Series headtops
  • Optional shroud for use with Versaflo™ M-407SG helmets featuring L-153SG Speedglas™ attachment
  • An M-444 Inner Shroud (sold separately) must be zipped into the outer shroud.
  • 3M Catalog# M-449

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI Z89.1 Type I, Class G
Color Black
Disposable Reusable
Material Leather
Product Type Leather Shroud
Size Standard
For Use With (Application) Welding
For Use With (Equipment) M-Series Helmet Assembly
Model M-449
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.