Learn More
MSC VERSAFLO PAPER ASSEMBLY
Catalog No. 5000674958
Encompass
The 3M Belt Mounted PAPR Motor Blower Unit is a heavy-duty assembly designed for powered air-purifying respirators. It includes a rechargeable lithium-ion battery and is part of the TR-300 Series.
  • PAPR unit blower type
  • Belt mount type
  • Lithium-ion battery chemistry
  • Includes battery, belt, and TR-306N+ PAPR unit
  • Not intrinsically safe
  • Not flame resistant
Catalog No. 50-006-74958 Supplier MSC Supplier No. 46721627
The 3M Belt Mounted PAPR Motor Blower Unit is a heavy-duty assembly designed for powered air-purifying respirators. It includes a rechargeable lithium-ion battery and is part of the TR-300 Series.
  • PAPR unit blower type
  • Belt mount type
  • Lithium-ion battery chemistry
  • Includes battery, belt, and TR-306N+ PAPR unit
  • Not intrinsically safe
  • Not flame resistant