Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ PAPR Assembly TR-304N+

Catalog No. 19039578 Shop All 3M Products
Change view
Click to view available options
Beige
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-039-578 Beige
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-039-578 Supplier 3M™ Supplier No. 7100153811/DEL

missing translation for 'validMainframeMsg'

PAPR provides protection against particulates. Combines performance, protection and functionality with a slim, lightweight balanced design for comfort.

  • Two selectable flow rate options - 6.5 CFM and 7.2 CFM - offers more options and comfort to the user
  • Easy-to-use and versatile PAPR system for particulate environments
  • High Efficiency particulate filter
  • Economy battery included
TRUSTED_SUSTAINABILITY

Specifications

Battery Life 6 hours
Battery Type Economy Battery
Certifications/Compliance NIOSH Approved
Color Beige
Filter Type High Efficiency (HE)
Product Type PAPR Assembly, No Headtop
For Use With (Application) Powder Handling
Includes 3M™ Versaflo™ PAPR Assembly, Easy Clean Belt, Economy Battery
Model TR-304N
Mounting Type Belt-mounted
Quantity 1 Ea.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.