Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ PAPR Assembly TR-614-HK

Catalog No. 19121503 Shop All 3M Products
Change view
Click to view available options
Color: Gray
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-121-503 Color: Gray
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-121-503 Supplier 3M™ Supplier No. 7100098537
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

3M™ Versaflo™ PAPR Assembly TR-614-HK allows workers to customize thier respiratory protection system by choosing the headtop.

  • Utilizes cartridges to protect against gas and organic vapors, combined with filters for protection against particulates
  • Longer run time, reduced downtime with a high capacity battery and charge-level indicators to keep you prepared and informed
  • See more of your work world with this convenient, maintenance-free disposable headcover
  • Fully submersible product features a smooth surface design that’s quick and easy to clean and decontaminate
  • 3M Catalog# TR-614-HK

TRUSTED_SUSTAINABILITY

Specifications

Battery Life 4 to 12 hours
Battery Type Lithium-ion
Cartridge Type Gas and Organic Vapors (sold separately)
Certifications/Compliance NIOSH Approved
Color Gray
Filter Type OV/HE Filter
Product Type PAPR Assembly, No Headtop
For Use With (Application) Health and Safety Testing
Model TR-614-HK
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.