Learn More
3M™ Versaflo™ PAPR Breathing Tube Covers
Catalog No. p-7225021 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
TR-300+ PAPR
TR-600/800 PAPR
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
19-807-976 TR-300+ PAPR
19-807-975 TR-600/800 PAPR
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-807-976 Supplier 3M™ Supplier No. 7100260719
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
PAPR disposable breathing covers are designed to help keep Versaflo™ PAPRs clean from debris.
  • Made from a liquid resistant material with zippers for convenient donning and doffing
  • Clear plastic window for control viewing and button activation
  • Helps keep PAPR clean from debris
  • Compatible with Versaflo™ PAPRs
  • Popular for paint spray application
  • Made with no natural rubber latex components
TRUSTED_SUSTAINABILITY
Specifications
Material Plastic
Product Type Breathing Tube Cover
For Use With (Application) Protects PAPR blower from dirt and debris
For Use With (Equipment) TR-300+ PAPR
Model TR-381