Learn More
MSC Versaflo PAPR, Easy Clean Assembly, Intrinsically Safe, 3M, 1 ea
Catalog No. 5000673755
Encompass
This Versaflo Powered Air Purifying Respirator Easy Clean Assembly, part of the TR-800 Series, is a NIOSH-approved, intrinsically safe motor blower unit designed for hazardous locations. It provides respiratory protection in environments with flammable gases or combustible dusts.
  • Intrinsically safe for hazardous locations (Exia Division 1)
  • NIOSH-approved powered air purifying respirator system
  • Rechargeable lithium-ion battery
  • Belt-mounted design
  • Audible and visual alarms for low battery and low airflow
  • Compatible with loose-fitting headgear
  • Supports particulate-only filters or combination particulate and gas/vapor cartridges
Catalog No. 50-006-73755 Supplier MSC Supplier No. 46721601
This Versaflo Powered Air Purifying Respirator Easy Clean Assembly, part of the TR-800 Series, is a NIOSH-approved, intrinsically safe motor blower unit designed for hazardous locations. It provides respiratory protection in environments with flammable gases or combustible dusts.
  • Intrinsically safe for hazardous locations (Exia Division 1)
  • NIOSH-approved powered air purifying respirator system
  • Rechargeable lithium-ion battery
  • Belt-mounted design
  • Audible and visual alarms for low battery and low airflow
  • Compatible with loose-fitting headgear
  • Supports particulate-only filters or combination particulate and gas/vapor cartridges