Learn More
3M™ Versaflo™ PAPR Rechargeable Battery Power Supply Devices
Catalog No. 19268025 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Breathe Easy PAPR Systems
TR-630 and TR-632 Batteries
Versaflo TR-300 PAPRs
Versaflo TR-300 or Speedglas TR-300-SG PAPRs
Versaflo TR-300+, TR-600, TR-800 PAPRs
Versaflo TR-300, TR-300+, TR-600, TR-800 PAPRs
Product Type:
Battery Charger
Multi-Unit Charging Station
Single-Unit Charging Station
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
19-268-025 TR-630 and TR-632 Batteries Multi-Unit Charging Station
19-039-568 Versaflo TR-300 or Speedglas TR-300-SG PAPRs Multi-Unit Charging Station
19-268-024 - Single-Unit Charging Station
19-039-575 Versaflo TR-300, TR-300+, TR-600, TR-800 PAPRs Battery Charger
19-039-576 Versaflo TR-300+, TR-600, TR-800 PAPRs Multi-Unit Charging Station
19-039-567 Versaflo TR-300 PAPRs Single-Unit Charging Station
18-783B Breathe Easy PAPR Systems Multi-Unit Charging Station
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 19-268-025 Supplier 3M™ Supplier No. 7100035839
Choice of single or multiple unit power supply sources
  • Provide extended use of powered air purifying respirator systems
  • 3M Catalog# GVP-110, GVP-210, TR-341N, TR-344N, TR-641N, TR-644N, TR-941N, TR-944N, 520-04-24
Compliance
NIOSH approved.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Multi-Unit Charging Station
For Use With (Equipment) TR-630 and TR-632 Batteries
No. of Ports Four