Learn More
MSC Versaflo Peel-Off Visor Cover, 3M, 4-Layer, S-Series, 10 EA/Case
Catalog No. 5000686550
Encompass
This multi-layer transparent visor cover is designed for S-Series hood assemblies to protect the visor from paint overspray, contaminants, and scratches. It consists of four peel-off layers in a preassembled stack.
  • Four layer peel-off film helps protect the visor from contaminants and scratches
  • Multi-layer peel-off extends the life of the hood in paint spray applications
  • Full perimeter adhesive enables easy attachment to the visor
  • Optimized for S-series hood assemblies
Catalog No. 50-006-86550 Supplier MSC Supplier No. 66994922
This multi-layer transparent visor cover is designed for S-Series hood assemblies to protect the visor from paint overspray, contaminants, and scratches. It consists of four peel-off layers in a preassembled stack.
  • Four layer peel-off film helps protect the visor from contaminants and scratches
  • Multi-layer peel-off extends the life of the hood in paint spray applications
  • Full perimeter adhesive enables easy attachment to the visor
  • Optimized for S-series hood assemblies