Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3M™ Versaflo™ Powered Air Purifying Respirator PAPR Assembly TR-612N

Catalog No. 19070962 Shop All 3M Products
Change view
Click to view available options
Color: Black
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
19-070-962 Color: Black
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-070-962 Supplier 3M™ Supplier No. 7100176754
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

3M™ Versaflo™ Powered Air Purifying Respirator PAPR Assembly TR-612N can be combined with 3M Full Facepiece Reusable Respirators.

  • Easy to Use
  • Easy to buy assembly of commonly used components
  • Light weight
  • Easy to clean
  • Easy to maintain
  • Customize the system for your environment
  • Compatible 3M tight fitting full facepiece respirators
  • Choose between filters and/or a wide range of chemical cartridges
  • Fully submersible for cleaning when using cleaning and storage plugs
  • 3M Catalog# TR-612N

TRUSTED_SUSTAINABILITY

Specifications

Battery Life 8 hours
Battery Type High Capacity Lithium Ion
Certifications/Compliance NIOSH Approved
Color Black
Product Type PAPR Assembly, No Headtop
For Use With (Application) General Maintenance, Liquid Chemical Handling, Machine Operations, Spray Insulation, Demolition
For Use With (Equipment) 3M™ Versaflo ™ Powered Air Purifying Respirators
Includes Motor/Blower, Standard Battery and Charger, Heavy Duty Breathing Tube, High Durability Belt, Airflow Indicator, Sspark Arrestor Pre-filter, Pre-filter, 2 Caps and Storage Plug
Model TR-612N
Quantity 1 Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.